Order Entry
Slovenia
ContactUsLinkComponent
Anti-P27 KIP 1 Polyclonal Antibody
Anti-P27 KIP 1 Polyclonal Antibody
Catalog # BSBTPB9070
Supplier:  Boster Bio
undefined
Anti-P27 KIP 1 Polyclonal Antibody
Catalog # BSBTPB9070
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    P27 KIP 1
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Polyclonal antibody for p27/CDKN1B detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. p27/CDKN1B information: Molecular Weight: 22073 MW; Subcellular Localization: Nucleus. Cytoplasm. Endosome . Nuclear and cytoplasmic in quiescent cells. AKT- or RSK- mediated phosphorylation on Thr-198, binds 14-3-3, translocates to the cytoplasm and promotes cell cycle progression. Mitogen- activated UHMK1 phosphorylation on Ser-10 also results in translocation to the cytoplasm and cell cycle progression. Phosphorylation on Ser-10 facilitates nuclear export. Translocates to the nucleus on phosphorylation of Tyr-88 and Tyr-89. Colocalizes at the endosome with SNX6; this leads to lysosomal degradation (By similarity); Tissue Specificity: Expressed in all tissues tested. Highest levels in skeletal muscle, lowest in liver and kidney.

Type: Primary
Antigen: P27 KIP 1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Rat