Acetic acid 80% ≥80% (v/v) in water
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid
CAS Number:64-19-7
:
Formula:H₃CCOOH
MDL Number:MFCD00036152
Boiling Pt:118
Molecular Weight:60.05 g/mol
Melting Pt:17
Flash Pt:>60 °C
Storage Temperature:Ambient
UN:2790
ADR:8 II
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless Solution
- Density:1.067 to 1.071 kg/l
- Titration with NaOH:79.0 to 81.0 % v/v
Specification Test Results
Frequently Bought Together