Prilocaine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: N-(2-Methylphenyl)-2-(propylamino)propanamide
CAS Number:721-50-6
:
Formula:C₁₃H₂₀N₂O
MDL Number:MFCD00048681
Molecular Weight:220.31 g/mol
Melting Pt:36…38.7 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Melting point:36 to 39 °C
- Water:≤0.5 %
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together