Spray chamber
Catalog # HEWLG3139-65003
Supplier: Agilent
Specifications
- Description:Spray chamber
- Environmentally Preferable:
- Pk:1
Specifications
About this item
This PFA spray chamber is the standard spray chamber used with the 7500 ICP-MS when aspirating hydrofluoric acid.