CHO cell culture platform liquid media, HyClone™ HyCell™ TransFx-C
HyCell™ TransFx-C Media is a hydrolysate-free and regulatory-friendly media which supports transient transfection, growth, and expression of a variety of recombinant proteins in CHO cells.
- Defined hydrolosate-free ADCF cell culture media formulation
- Supports the transient expression of a variety of recombinant proteins
- Same media for transient transfection and production
- Developed through our Metabolic Pathway Design process
- Component traceability
High-performing, animal-derived component-free (ADCF) cell culture media with sodium bicarbonate and without L-glutamine and poloxamer 188.
: Manufactured in cGMP (21 CFR 820) compliant and ISO 9001 certified facilities.
- Environmentally Preferable:
Frequently Bought Together