HypoThermosol® biopreservation media is a novel, engineered, optimised hypothermic storage and shipping media product. Serum-free, protein-free HypoThermosol® is designed to provide maximum storage and shipping stability for biologics at 2 to 8 °C.
This proprietary, optimised formulation mitigates temperature-induced molecular cell stress responses that occur during chilling and re-warming of biologics, intermediate products, and final cell products intended for research and clinical applications.
HypoThermosol® FRS includes components that scavenge free radicals, provide pH buffering, oncotic/osmotic support, energy substrates, and ionic concentrations that balance the intracellular state at low temperatures.
This reducing post-preservation necrosis and apoptosis as compared to commercial and home-brew isotonic and extracellular formulations. This results in greatly extended shelf life and improved post-preservation viability.
- Source:
- Species:
- Environmentally Preferable: