- Antigen name:Enolase 1, (alpha)
- Antigen symbol:ENO1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Chicken
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:MBP-1|Phosphopyruvate hydratase|ENO1L1|NNE|Non-neural enolase|Alpha-enolase|Enolase 1|MBPB1|C-myc promoter-binding protein|ENO1|Plasminogen-binding protein|2-phospho-D-glycerate hydro-lyase|MPB1|MPB-1
- Concentration:1 mg/mL
- Immunogen:161-320
- Size:50 µg
- Pk:50 µG
Enolase 1; phosphopyruvate hydratase; MYC promoter-binding protein 1; non-neural enolase; 2-phospho-D-glycerate hydro-lyase; tau-crystallin. Enolase 1 gene encodes one of three enolase isoenzymes found in mammals; it encodes beta-enolase, a homodimeric soluble enzyme, and also encodes a shorter monomeric structural lens protein, tau-crystallin. The two proteins are made from the same message. The full length protein, the isoenzyme, is found in the cytoplasm. The shorter protein is produced from an alternative translation start, is localized to the nucleus, and has been found to bind to an element in the c-myc promoter.
NNE antibody can be used for the detection of NNE by Western blot, may also work for IHC and ICC.
Antigen affinity-purified
Phosphate-Buffered Saline. No preservatives added.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ENO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat