Order Entry
Slovenia
ContactUsLinkComponent
 
Anti-ERAP2 Rabbit Polyclonal Antibody
Catalog # PRSI7469
Supplier:  ProSci Inc.
undefined
Anti-ERAP2 Rabbit Polyclonal Antibody
Catalog # PRSI7469
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Endoplasmic reticulum aminopeptidase 2
  • Antigen symbol:
    ERAP2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Liquid
  • Antigen synonyms:
    LRAP|Endoplasmic reticulum aminopeptidase 2|L-RAP|Leukocyte-derived arginine aminopeptidase|ERAP2
  • Concentration:
    1 mg/mL
  • Immunogen:
    ERAP2 antibody was raised against a 17 amino acid peptide near the amino terminus of human ERAP2.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

The endoplasmic reticulum (ER) aminopeptidase 2 (ERAP2), a member of the peptidase M1 family, like the related protein ERAP1, plays a central role in peptide trimming, a step required for the generation of most HLA class I-binding peptides (1,2). Like ERAP1, ERAP2 is localized to the lumen of the ER and is thought to associate with ERAP1 as a heterodimer. Both ERAP1 and ERAP2 have been linked to several human diseases ranging from infections to autoimmunity and cancer, and may play a role in the innate immune response .

ERAP2 Antibody is affinity chromatography purified via peptide column.

ERAP2 Antibody is supplied in PBS containing 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ERAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human