Specifications
- Antigen name:Death-associated Protein
- Antigen symbol:DAP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:DAP1|DAP-1|DAP|Death-associated protein 1
- Concentration:1 mg/mL
- Immunogen:DAP antibody was raised against a 19 amino acid peptide near the carboxy terminus of human DAP.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
DAP Antibody: The Death-associated protein (DAP) is a basic proline-rich 15kDa protein that as a positive mediator of programmed cell death that is induced by interferon-gamma. DAP is also a direct substrate of mammalian target of rapamycin (mTOR), a serine/threonine kinase that regulates cell growth and cell cycle, and a negative regulator of autophagy. Under rich nutrient conditions, mTOR phosphorylates DAP at Ser3 and Ser51; under starvation conditions, these residues are dephosphorylated and DAP is converted into an active suppressor of autophagy.
DAP Antibody is affinity chromatography purified via peptide column.
DAP Antibody is supplied in PBS containing 0.02% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: DAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat