Anti-ARL2 Rabbit Polyclonal Antibody
Catalog # PRSI71-317
Supplier: ProSci Inc.
Specifications
- Antigen name:ADP-Ribosylation Factor-Like 2
- Antigen symbol:ARL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Antigen synonyms:ARFL2
- Concentration:1 mg/mL
- Immunogen:Synthetic peptide containing a sequence corresponding to a region within amino acids 123 and 184 of Human ARL2.
- Tested applications:Immunoaffinity Chromatography
- Size:50 µl
- Pk:50 µl
Specifications
About this item
ARL2 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 10 μg/mL, and Western Blotting starting at 1:500 - 1:3000.
PBS, pH 7, 1% BSA, 20% Glycerol, 0.01% Thimerosal
PBS, pH 7, 1% BSA, 20% Glycerol, 0.01% Thimerosal
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ARL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human