Anti-ATP13A1 Goat Polyclonal Antibody
Catalog # PRSI46-629
Supplier: ProSci Inc.
Specifications
- Antigen name:ATPase type 13A1
- Antigen symbol:ATP13A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:ATP13A1
- Concentration:500 ug/mL
- Immunogen:ATP13A1 antibody was raised against a 13 amino acid synthetic peptide near the internal region of ATP13A1.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 110 kDa band observed in human kidney lysates (calculated MW of 133 kDa according to NP_065143.2). Recommended concentration: 0.3-1 μg/mL.
ATP13A1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
ATP13A1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ATP13A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human