Anti-FHL1 Goat Polyclonal Antibody
Catalog # PRSI45-591
Supplier: ProSci Inc.
Specifications
- Antigen name:Four and a Half LIM Domains 1
- Antigen symbol:FHL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:FHL1A|LIM protein SLIMMER|XMPMA|KYO-T|MGC111107|RP11-535K18.1|SLIMMER|skeletal muscle LIM-protein1|four-and-a-half Lin11|KYOT|FHL1|SLIM1|Four-and-a-half LIM domains 1|four and a half LIM domains 1|Isl-1 and Mec-3 domains 1|FHL1B
- Concentration:500 ug/mL
- Immunogen:FHL1 antibody was raised against a 13 amino acid synthetic peptide near the internal region of FHL1.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
FHL1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
FHL1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: FHL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse