Anti-GAD1 Goat Polyclonal Antibody
PRSI42-864
- Antigen name:Glutamate Decarboxylase 1 (brain 67kDa)
- Antigen symbol:GAD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:67kDa)|SCP|glutamate decarboxylase 1|67kD)|glutamate decarboxylase 1 (brain|CPSQ1|GAD|GAD1|OTTHUMP00000041055|GAD67|GAD25|FLJ45882
- Concentration:500 ug/mL
- Immunogen:GAD1 antibody was raised against an 11 amino acid synthetic peptide near the internal region of GAD1.
- Size:100 µg
- Pk:100 µG
Antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
GAD1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: GAD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse