Anti-CHML Rabbit Polyclonal Antibody
Catalog # PRSI31-091
Supplier: ProSci Inc.
Specifications
- Antigen name:Choroideremia-like (Rab escort protein 2)
- Antigen symbol:CHML
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:Rab escort protein 2|Rab proteins geranylgeranyltransferase component A 2|Choroideremia-like protein|REP-2|REP2
- Immunogen:A synthetic peptide corresponding a region of human CHML.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
The product of the CHML gene supports geranylgeranylation of most Rab proteins and may substitute for REP-1 in tissues other than retina. CHML is localized close to the gene for Usher syndrome type II.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: CHML
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: