Anti-GBP4 Rabbit Polyclonal Antibody
Catalog # PRSI30-820
Supplier: ProSci Inc.
Specifications
- Antigen name:Guanylate Binding Protein 4
- Antigen symbol:GBP4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:Mpa2
- Immunogen:A synthetic peptide corresponding a region of human GBP4.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
GBP4 belongs to the GBP family. It binds GTP, GDP and GMP. Hydrolyzes GTP very efficiently; GDP rather than GMP is the major reaction product. GBP4 plays a role in erythroid differentiation.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: GBP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human