- Antigen name:Influenza Virus NS1A Binding Protein
- Antigen symbol:NS1BP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Dog
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:KLHL39|HSPC068|NS-1|NS1BP|ND1|NS1-BP|FLARA3
- Immunogen:A synthetic peptide corresponding a region of human IVNS1ABP.
- Size:50 µg
- Pk:50 µG
This gene encodes a protein which interacts with the nonstructural NS1 protein of the influenza A virus. In noninfected cells, affinity-purified antibodies localized this protein in nuclear regions enriched with the spliceosome assembly factor SC35, suggesting an association with the cellular splicing apparatus. In influenza A virus-infected cells, the protein relocalized throughout the nucleoplasm and appeared distinct from the SC35 domains, which suggests that its function may be disturbed or altered.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: NS1BP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human