About this item
ProClin™ are among the most widely used presevatives in the in vitro diagnostic (IVD) industry. These materials are water-soluble formulations that are easily used directly from the container. They have no health hazards, toxicology problems, or disposal issues at recommended usage levels.
- Compatibility with key enzymes and diagnostic assay indicators
- Will not inhibit antibody binding
- Excellent stability over a wide pH range
- Stable in TRIS, TES and HEPES buffer up to a pH of 7
- Level of active ingredients assayable by HPLC
- Safe and easy to use, water soluble liquid
ProClin™ presevatives are immediately microstatic upon contact with microbial organisms as a result of their ability to quickly penetrate cell membranes and inhibit specific enzymes crucial to cellular respiration. Multiple specific sites of ProClin™ toxicity prevent microorganisms from developing high levels of resistance, making ProClin™ highly efficient even over extended storage periods of your IVD components.
Appearance: Clear, colorless to light yellow liquid, free of foreign matter.
Specifications
- Catalog No:
- SIAL48912-U
- SIAL48914-U
- SIAL48917-U
- SIAL48918-U
- SIAL48915-U
- M-Clarity quality level:
- MQ300
- MQ300
- MQ300
- MQ300
- MQ300