Cleanroom tables, ergo
Supplier: Palbam
Certificates
About this item
These cleanroom ergo tables are manufactured with 304 stainless steel with electropolished finish and Al anodized cleanroom compatible connectors.
- Rounded front table tops
- Excellent design conforms to highest cleanroom specifications
- Ergonomic table legs designed to prevent operators 'knocking knees' on table
- Height adjustable feet or casters
- All accessories attachable
The bases are fabricated from tubing for easy cleaning and optimal airflow. The tops are designed to reduce vibrations and are very strong holding up to 80 kg.
Delivery: The tables are shipped flat-packed with easy assembly instructions (assembly time ~10 minutes). Other shelves are available upon request.
Frequently Bought Together