Horizontal gel electrophoresis units, Axygen®
: Corning
Axygen® horizontal gel box is a high quality, full-featured unit designed for exceptional durability and efficiency. These gel boxes feature leak-free external gel casting, which allows the user to cast one gel outside of the box while another gel is running inside. Boxes from 7, 10, 15 and 20cm
- Flexibility – Each unit comes with 2 sizes of gel trays to optimize applications
- External gel casting to gel box allows for running gels while casting the next one
- Ease of use – the Axygen contrast enhancer makes loading gels easier
- Rugged injection molding is supported by a 3-year warranty
- Larger units come with recirculation ports allowing for buffer exchange
- Catalog No:
- AXYGHGB-7
- AXYGHGB-20
- AXYGHGB-15
- AXYGHGB-10
- Model:
- HGB-7
- HGB-10
- Tank volume:
- 1500 ml (buffer volume)
- 500 ml (buffer volume)
- 300 ml (buffer volume)
- Width:
- 23 cm
- 17,5 cm
- 12.5 cm
- Length:
- 39.5 cm
- 26.5 cm
Frequently Bought Together