Anti-KLHL9 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
BOSSBS-7753R-HRP
: Bioss
- Antibody type:Primary
- Antigen name:Kelch-like 9
- Antigen symbol:KLHL9
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:55958
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
KLHL9 is the substrate-specific adapter for a CUL3-based E3 ubiquitin-protein ligase complex. Within this complex, KLHL9 controls the dynamic behavior of AURKB on mitotic chromosomes and thereby coordinates faithful mitotic progression and completion of cytokinesis.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: KLHL9
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat