Anti-CAMK4 Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-6728R-CY5.5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Calcium/calmodulin-dependent Protein Kinase 4
- Antigen symbol:CAMK4
- Clonality:Polyclonal
- Conjugation:Cy5.5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:CAMK|CaMK IV|CAMK-GR|CAMK4|Calcium/calmodulin-dependent protein kinase type IV|CAMKIV|CaM kinase-GR
- Modification:phospho-Thr196/Thr200
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:192-203/469
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Ca2+/calmodulin dependent protein kinase type IV (CAMKIV) belongs to the serine/threonine protein kinase family, and to the Ca(2+)/calmodulin-dependent protein kinase subfamily. This enzyme is a multifunctional serine/threonine protein kinase with limited tissue distribution, that has been implicated in transcriptional regulation in lymphocytes, neurons and male germ cells.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CAMK4
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat