Specifications
- Antibody type:Primary
- Antigen name:Protein Kinase C Substrate 80K-H
- Antigen symbol:PRKCSH
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:5589
- Antigen synonyms:AGE-R2|PLD1|PKCSH|G19P1|PCLD
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Trimming of glucoses from N-linked core glycans on newly synthesized glycoproteins occurs sequentially through the action of Glucosidases I and II in the endoplasmic reticulum (ER). Glucosidase II is an ER-localized enzyme that contains a and b subunits (Glucosidase IIa and Glucosidase IIb) which form a defined heterodimeric complex. Glucosidase IIa is the catalyitc core of the enzyme and can function independently of the b subunit. The sequence of Glucosidase IIb encodes protein rich in glutamic and aspartic acid with a putative ER retention signal (HDEL) at the C-terminus. The phosphorylated form of Glucosidase IIb is localized in the plasma membrane and is highly expressed in FGF-stimulated fibroblasts and epidermal carcinoma cells. Glucosidase IIb was first purified from a human carcinoma cell line as a potential substrate for protein kinase C. Through the HDEL signal at the C-terminus, Glucosidase IIb retains the complete complex in the ER.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PRKCSH
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat