Anti-NOX4 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
BOSSBS-1091R-FITC
: Bioss
- Antibody type:Primary
- Antigen name:NAPDH oxidase 4
- Antigen symbol:NOX4
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Dog,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:50490
- Antigen synonyms:NAPDH oxidase 4|KOX-1|KOX|NOX-4|RENOX
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:84-105/578
- Purification:Purified by Protein A
- Pk:100 µl
Constitutive NADPH oxidase which generates superoxide intracellularly upon formation of a complex with CYBA/p22phox. Regulates signaling cascades probably through phosphatases inhibition. May function as an oxygen sensor regulating the KCNK3/TASK-1 potassium channel and HIF1A activity. May regulate insulin signaling cascade. May play a role in apoptosis, bone resorption and lipolysaccharide-mediated activation of NFKB.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NOX4
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog