Anti-ABCC1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-0657R-A488
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:ATP-binding Cassette, Subfamily C (CFTR/MRP), Member 1
- Antigen symbol:ABCC1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:17250
- Antigen synonyms:multiple drug resistance-associated protein|ABC29|DKFZp781G125|DKFZp686N04233|MRP1|LTC4 transporter|multidrug resistance protein|ABCC|sub-family C (CFTR/MRP)|sub-family C|member 1|GS-X|ATP-binding cassette|leukotriene C(4) transporter|multiple drug resistance protein 1|MRP|ABCC1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:500-548/1528
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Mediates export of organic anions and drugs from the cytoplasm. Mediates ATP-dependent transport of glutathione and glutathione conjugates, leukotriene C4, estradiol-17-beta-o-glucuronide, methotrexate, antiviral drugs and other xenobiotics. Confers resistance to anticancer drugs. Hydrolyzes ATP with low efficiency.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ABCC1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat