Anti-p-ErbB2/HER2 Rabbit Polyclonal Antibody [clone: Y877]
Catalog # BWRLBS4091
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-ErbB2/HER2
- Clonality:Polyclonal
- Clone:Y877
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human ErbB2/HER2 around the phosphorylation site of Tyrosine 877.
p-ErbB2/HER2 (Y877) pAb detects endogenous levels of ErbB2/HER2 protein only when phosphorylated at Tyr877.
Type: Primary
Antigen: p-ErbB2/HER2
Clonality: Polyclonal
Clone: Y877
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat