Order Entry
Slovenia
ContactUsLinkComponent
Anti-PSK-H1 Rabbit Polyclonal Antibody [clone: M295]
Anti-PSK-H1 Rabbit Polyclonal Antibody [clone: M295]
  BWRLBS2256
Anti-PSK-H1 Rabbit Polyclonal Antibody [clone: M295]
  BWRLBS2256

 

  • Antibody type:
    Primary
  • Antigen symbol:
    PSK-H1
  • Clonality:
    Polyclonal
  • Clone:
    M295
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Synthetic peptide, corresponding to amino acids 261-310 of Human PSK-H1.

PSK-H1 (M295) pAb detects endogenous levels of PSK-H1 protein.

Type: Primary
Antigen: PSK-H1
Clonality: Polyclonal
Clone: M295
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse