Anti-MEK-4 Rabbit Polyclonal Antibody [clone: E74]
BWRLBS1320
- Antibody type:Primary
- Antigen symbol:MEK-4
- Clonality:Polyclonal
- Clone:E74
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 50-100 of Human MEK-4.
MEK-4 (E74) pAb detects endogenous levels of MEK-4 protein.
Type: Primary
Antigen: MEK-4
Clonality: Polyclonal
Clone: E74
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat