Contact Plates with Locking-Lid Plate
Supplier: PHARMAMEDIA DR MULLER
New Product
Certificates
About this item
Used in pharmaceutical industry in clean rooms and isolators for contact sampling, personnel monitoring, as well as active air monitoring. Isolation and growth of fastidious bacteria, yeasts and moulds incubations in vent and closed position possible. Easy handling due to increased handling area.
- Detection of aerobic and anaerobic micro-organisms
- Gamma irradiated and triple-bagged
- Locking-lid plate, made from polystyrene guarantees save transport
- Label on side of bottom
- High barrier foil for H₂O₂ as well as for water vapour
Certifications: According Ph. Eur. and Pharmacopoeia.
Delivery: All plates are delivered in the non-locked position, and will be delivered refrigerated.
Packaging: 10 plates per packaging (6 or 10 staples) unit.
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together