Anti-EGFR Mouse Monoclonal Antibody [clone: GFR/1667]
Catalog # ABNOMAB14510
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Epidermal growth factor receptor Recombinant
- Antigen symbol:EGFR
- Clonality:Monoclonal
- Clone:GFR/1667
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Environmentally Preferable:
- Form:Liquid
- Gene ID:1956
- Storage buffer:In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
- Storage temperature:Store at 4° C.
- Immunogen:Recombinant protein corresponding to full length human EGFR.
- Purification:Protein A/G purification
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against full length recombinant human EGFR.
Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone: GFR/1667
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human