Order Entry
Slovenia
ContactUsLinkComponent
Bordetella pertussis toxin
Bordetella pertussis toxin
Catalog # ENZOBMLG1010050
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
Bordetella pertussis toxin
Catalog # ENZOBMLG1010050
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
Specifications
  • Pk:
    50 µG
  • Description:
    Bordetella pertussis toxin, 50 µg
  • Seq:
    02
  • Environmentally Preferable:
Specifications
About this item
Bordetella pertussis toxin (Islet-activating protein, PTX) from Bordetella pertussis, strain NCTC 10739. is a high purity toxin applicable to ELISA, antibody capture, cytotoxicity studies, LINE assay.
  • High purity, native protein available as >98% pure
  • Minimal lot-to-lot variation ensures reproducible results
  • Lyophilised format for extended shelf-life