Anti-NAPSA Mouse Monoclonal Antibody (CF640R) [clone: NAPSA/1238]
Supplier: Biotium
Certificates
About this item
Napsin is a pepsin-like aspartic proteinase connected with maturation of surfactant protein B.There are two closely related napsins, napsin A and napsin B. Napsin A is expressed as a single chain protein. Immunohistochemical studies revealed high expression levels of napsin A in human lung and kidney but low expression in spleen. Napsin A is expressed in type II pneumocytes and in adenocarcinomas of lung. The high specificity expression of napsin A in adenocarcinomas of lung is useful to distinguish primary lung adenocarcinomas from adenocarcinomas of other organs.
Type: Primary
Antigen: NAPSA
Clonality: Monoclonal
Clone: NAPSA/1238
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Specifications
- Catalog No:
- BNC401238-500
- BNC401238-100
- Antigen symbol:
- NAPSA
- NAPSA
- Antigen name:
- Napsin A
- Napsin A
- Antigen synonyms:
- NAPA|Aspartyl protease 4|Kidney derived aspartic protease like protein (Kdap)|KAP|Pronapsin A|NAP1|Napsa|SNAPA|napsin A aspartic peptidase|ASP4
- NAPA|Aspartyl protease 4|Kidney derived aspartic protease like protein (Kdap)|KAP|Pronapsin A|NAP1|Napsa|SNAPA|napsin A aspartic peptidase|ASP4
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 640R
- CF® 640R
- Clone:
- Clone NAPSA/1238
- Clone NAPSA/1238
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 512843
- 512843
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
Specifications
Frequently Bought Together