Order Entry
Slovenia
ContactUsLinkComponent
Anti-MYADM Mouse Monoclonal Antibody (Purified) [clone: MYADM/972]
Anti-MYADM Mouse Monoclonal Antibody (Purified) [clone: MYADM/972]
Supplier:  Biotium
Certificates
Anti-MYADM Mouse Monoclonal Antibody (Purified) [clone: MYADM/972]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNUB0972-500
  • BNUB0972-100
  • Antigen symbol:
  • MYADM
  • MYADM
  • Antigen name:
  • Myeloid-associated differentiation marker
  • Myeloid-associated differentiation marker
  • Antigen synonyms:
  • Myeloid upregulated protein|myeloid associated differentiation marker|MYADM|Protein SB135
  • Myeloid upregulated protein|myeloid associated differentiation marker|MYADM|Protein SB135
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • Clone MYADM/972
  • Clone MYADM/972
  • Reactivity:
  • Macaque, Human
  • Macaque, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 380906
  • 380906
  • Isotype:
  • IgG1
  • IgG1
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,2 mg/ml
  • 0,2 mg/ml
  • Storage buffer:
  • PBS/0,05% BSA/0,05% azide
  • PBS/0,05% BSA/0,05% azide

Specifications

Frequently Bought Together