Anti-CAPON Rabbit Polyclonal Antibody
BOSSBS-11987R
: Bioss
- Antibody type:Primary
- Antigen name:C terminal PDZ domain ligand of neuronal nitric oxide synthase CAPON
- Antigen symbol:CAPON
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:NOS1AP|MGC138500|C terminal PDZ ligand of neuronal nitric oxide synthase protein|Carboxyl terminal PDZ ligand of neuronal nitric oxide synthase protein|Ligand of neuronal nitric oxide synthase with carboxyl terminal PDZ domain|C-terminal PDZ ligand of neuronal nitric oxide synthase protein|CAPON|CAPON_HUMAN|Nitric oxide synthase 1 neuronal adaptor protein|Nitric oxide synthase 1 adaptor protein.|Carboxyl-terminal PDZ ligand of neuronal nitric oxide synthase protein|C terminal PDZ domain ligand of neuronal nitric oxide synthase CAPON|C terminal PDZ domain ligand of neuronal nitric oxide synthase
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A
- Size:100 μg
- Pk:100 µl
Type: Primary
Antigen: CAPON
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat