Order Entry
Slovenia
ContactUsLinkComponent
Pure quartz air sampling filters, grade QM-H, Whatman™
Pure quartz air sampling filters, grade QM-H, Whatman™
Certificates
undefined
Pure quartz air sampling filters, grade QM-H, Whatman™
Certificates
Specifications
  • Catalog No:
  • 513-0124
  • 513-0123
  • 513-0125
  • 513-0122
  • 513-0094
  • Grade:
  • QM-H
  • QM-H
  • QM-H
  • QM-H
  • QM-H
  • Weight:
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • Thickness:
  • 430 µm
  • 430 µm
  • 430 µm
  • 430 µm
  • 430 µm
  • Max. temperature:
  • 900 °C
  • 900 °C
  • 900 °C
  • 900 °C
  • 900 °C