emulsion 30% Simethicone USP
Catalog # DOWC117544-200
Supplier: DOW CORNING
CAS Number:
Specifications
- Pk:200 kg
- Seq:4676
- Environmentally Preferable:
Specifications
Specification Test Results
- Nonvolatile Content:41,7%
- pH:2,6
- Siloxane Content:30,4%
Specification Test Results
About this item
Synonyms: Anti-foaming agent
: