jetMESSENGER® is a mRNA transfection reagent that has been specifically designed to offer outstanding transfection efficiency in cells that are usually hard to transfect, allowing relevant gene expression experiments in all cell types.
- Unmatched transfection efficiency
- High efficiency on a wide variety of difficult to transfect cells
- Outperform DNA transfection by switching to mRNA
- Extremely gentle on cells
- No risk of genome integration
- Perfectly suited for CRISPR/Cas-9 gene editing, iPS generation, stem cell differentiation and immunotherapy assays
mRNA transfection is as easy as DNA transfection, with the advantage that mRNA does not need to reach the cell nucleus for expression and does not require cell division for efficient gene expression. Hence, cells that are slow dividing or that have developed specific mechanisms to protect their genome can be used for gene expression. Free of charge samples for evaluation or bulk quantities are available upon request, please contact your local Avantor sales office for more details.
jetMESSENGER® has been designed to transfect cells that divide slowly or that have developed specific mechanisms, such as primary cells, neurons, suspension cells and various cancer cell lines. jetMESSENGER® enables transfection of these so-called hard to transfect cells, as well as easy to transfect cell types. jetMESSENGER® is very gentle on cells enabling high cell viability to be maintained.
- Environmentally Preferable: