Order Entry
Slovenia
ContactUsLinkComponent
High voltage holiday detectors
undefined
High voltage holiday detectors
  • Catalog No:
  • SIMELD8504
  • SIMELD8503
  • Batteries:
  • Gel cell 3 Ah slide off
  • Gel cell 3 Ah slide off
  • Display:
  • LCD
  • LCD
  • W×D×H:
  • 260×160×70 mm
  • 260×160×70 mm
  • Weight:
  • 2200 g
  • 2200 g
Frequently Bought Together