Order Entry
Slovenia
ContactUsLinkComponent
Wedge for measuring reverse-side tolerance
Wedge for measuring reverse-side tolerance
Catalog # SIMESP6000
Supplier:  Industrial Physics
undefined
Wedge for measuring reverse-side tolerance
Catalog # SIMESP6000
Supplier:  Industrial Physics
Specifications
  • Description:
    Wedge for measuring reverse-side tolerance, range 1 - 10 mm
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
Anodised aluminum wedge for quick measurement of reverse-side tolerance of windows and doors.
  • Tolerance is given in millimetres
  • Also equipped with a measuring scale for optically determining the thickness or glass