Wall cleaning tool
Supplier: PFENNIG REINIGUNGSTECHNIK
Certificates
About this item
Wall cleaning tool for wall and ceiling cleaning in hygienically sensitve areas.
- Frame completely made of stainless steel
- Mops are 100% polyester microfiber filament yarn
- Autoclavable, disinfectant proof
- Mops have high-quality stainless steel push buttons for fitting with frame
This mop frame with locking screw is suitable with mop covers Micro Sicuro CR/A and Mop CR for wall cleaning tool.
Packaging: Mops are prewashed and single double packaged.
Frequently Bought Together