Test kits, photometric, iron, LCK cuvette tests
: Hach
Cuvette tests for water testing applications.
- Precise and reliable measurement values
- Maximum safety for users, thanks to the closed system and low amounts of reagents
- Convenient and error-free dosing of the reagents
- Barcode label for automatic recognition in the photometer
- Differentiation between tests and measuring ranges by means of colour-coding
Shelf life (LCK321): 30 months from production date
Shelf life (LCK320 and LCK521): 12 months from production date
Storage conditions: 2 to 8 °C (keep refrigerated)
: LCK321 and LCK521: ISO 6332-1988, DIN 38406 E1-1
: Cuvette tests are officially approved for many legally required consent limits, with the help of Addista standard solutions and round-robin test solutions, they provide the assurance you need.
- Environmentally Preferable:
Frequently Bought Together