(S)-(+)-2-Phenylglycine methyl ester hydrochloride 97%
: (S)-Methyl 2-amino-2-phenylacetate hydrochloride, H-Phg-OMe·HCl, L-(+)-2-Phenylglycine methyl ester hydrochloride
:15028-39-4
:
:C₉H₁₂ClNO₂
:MFCD00077158
:201.65 g/mol
:200 °C (dec.)
:GHS07
:Refrigerator
- Environmentally Preferable:
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤1 %
- HPLC:≥96.0 %
- Specific optical rotation:+131° to +137° (20°C, 589 nm) (c=1,MeOH)
Frequently Bought Together