Magnesium bromide, anhydrous 98%, pure
Supplier: Thermo Fisher Scientific
About this item
- CAS Number:7789-48-2
- Formula:Br₂Mg
- MDL Number:MFCD00011105
- Molecular Weight:184.11 g/mol
- Melting Pt:711 °C
- Density:3,72 g/cm³ (20 °C)
- :Density: 3.72 g/cm³ (20 °C)
- Storage Temperature:Ambient
2 Options Available Below
Product Details & Documents
- CAS Number:7789-48-2
- Formula:Br₂Mg
- MDL Number:MFCD00011105
- Molecular Weight:184.11 g/mol
- Melting Pt:711 °C
- Density:3,72 g/cm³ (20 °C)
- :Density: 3.72 g/cm³ (20 °C)
- Storage Temperature:Ambient
Documents
ACRO41339
SDS
Specifications for Product Family
Specification Test Results
- Appearance:White to grey Powder or chunks
- Titration Argentometric:97.5 to 102.5 %
- Refractive index:1.5680 to 1.5700 (20°C, 589 nm)
- GC:≥97.5 %



