Order Entry
Slovenia
ContactUsLinkComponent
 
Phenol : chloroform : iso-amyl alcohol (25:24:1) saturated with 100 mM Tris-EDTA to pH 8.0 stabilised for molecular biology DNase-, RNase-, protease-free pH 7.8-8.2
 
undefined
Phenol : chloroform : iso-amyl alcohol (25:24:1) saturated with 100 mM Tris-EDTA to pH 8.0 stabilised for molecular biology DNase-, RNase-, protease-free pH 7.8-8.2
 

 

  • Environmentally Preferable:

 

 

  • Appearance:
    Clear yellow to orange Solution
  • Stabilizer:
    0.08 to 0.12 % hydroxyquinoline
  • pH:
    7.8 to 8.2 (at 25°C)
  • Composition:
    mixture of Phenol/Chloroform/Isoamyl alcohol in the ratio of 25:24:1 (v/v/v)
  • Dnase,Rnase,Protease activity:
    None detected
  • Iron (Fe):
    ≤1 ppm
  • Heavy metals (as Pb):
    ≤1 ppm

 

 

Frequently Bought Together