meso-Hydrobenzoin 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: meso-1,2-Diphenylethylene glycol, meso-1,2-Diphenyl-1,2-ethanediol, rel-(1R,2S)-1,2-Diphenylethane-1,2-diol
CAS Number:579-43-1
:492-70-6
Formula:C₁₄H₁₄O₂
MDL Number:MFCD00064253
Molecular Weight:214.26 g/mol
Melting Pt:136…137 °C
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Crystalline powder and or lumps
- Infrared spectrum:Conforms
- Melting point:134 to 140 °C
- GC:≥94 %
Specification Test Results
Frequently Bought Together