2-Bromo-3-thiophenecarboxylic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 2-Bromothiophene-3-carboxylic acid
CAS Number:24287-95-4
:
Formula:C₅H₃BrO₂S
MDL Number:MFCD05865213
Molecular Weight:207.041 g/mol
Melting Pt:179…181 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to tan Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
- Titration with HClO4:≥97.5 %
Specification Test Results
Frequently Bought Together