Order Entry
Slovenia
ContactUsLinkComponent
Methyl-1-cyclohexene-1-carboxylate 98%
Methyl-1-cyclohexene-1-carboxylate 98%
Catalog # ACRO213710050
CAS Number:  
undefined
Methyl-1-cyclohexene-1-carboxylate 98%
Catalog # ACRO213710050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4750 to 1.4790 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specification Test Results

About this item

Synonyms: Methyl cyclohex-1-enecarboxylate
 :
 :140.182 g/mol
Storage Temperature:Ambient
CAS Number:18448-47-0
Formula:C₈H₁₂O₂
MDL Number:MFCD00001544
Boiling Pt:190…192 °C (760 mmHg)
Density:1.03 g/cm³
Flash Pt:73 °C