(1R,3R)-N-BOC-1-Aminocyclopentane-3-carboxylic acid 95%, 98% ee
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: (1R,3R)-3-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid
CAS Number:489446-85-7
:
Formula:C₁₁H₁₉NO₄
MDL Number:MFCD02259727
Molecular Weight:229.276 g/mol
Melting Pt:117.3 °C
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to light yellow Powder
- Infrared spectrum:Conforms
- GC:≥94 %
- Enantiomeric excess:≥97.5 %
Specification Test Results
Frequently Bought Together