1,3-Divinyltetramethyldisiloxane 97%
: Divinyltetramethyldisiloxane, Tetramethyl-1,3-divinyldisiloxane
:2627-95-4
:
:C₈H₁₈OSi₂
:MFCD00014933
:139 °C (760 mmHg)
:–99.7 °C
:0.813 g/cm³
:186.40 g/mol
:24 °C
:Ambient
:1993
:3 III
:GHS07
:GHS02
- Environmentally Preferable:
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4100 to 1.4130 (20°C, 589 nm)
- GC:≥96.0 %
Frequently Bought Together