VWR®, Absorbents - Mats, Rolls and Socks, for Oil, Water and Chemical Based Spills
VWR® absorbents and spill containment products help keep your employees and work environment safe from unexpected spills, leaks, drips or other accidents.
- Absorbs a variety of industrial liquids including oil, water, solvents and coolants
- Cannot be left outdoors as they will absorb water
- Maintenance and universal sorbents
For use with chemicals, oil and water.
Colour: Grey
Frequently Bought Together