Order Entry
Slovenia
ContactUsLinkComponent
Soap dispensers, Cleanse Washroom
Soap dispensers, Cleanse Washroom
Catalog # 115-2433
Supplier:  Deb-STOKO
undefined
Soap dispensers, Cleanse Washroom
Catalog # 115-2433
Supplier:  Deb-STOKO

Specifications

  • Capacity:
    2000 ml
  • Colour:
    White
  • Description:
    Soap dispenser, Cleanse Washroom for Refresh™ FOAM (DE, FR, NL)
  • Seq:
    02
  • Environmentally Preferable:
  • Pk:
    1

Specifications

About this item

The Cleanse Washroom are skincare dispensers for soaps and cleansers.

  • Wall-mounted durable plastic
  • For one-handed operation
  • Capacity 1000 ml or 2000 ml

For offices, medical rooms, laboratories, food services and catering with Refresh Azure FOAM, Refresh Clear FOAM, Refresh Energie FOAM, Refresh Rose FOAM.